Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMPRSS9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182973.1 | NP_892018.1 | 9 | 1462 | ||
XM_011527978.2 | XP_011526280.1 | 11 | 1825 | |||
XM_011527980.1 | XP_011526282.1 | 3 | 481 | |||
GenBank mRNA | AJ488946.1 | - | 9 | 1462 | ||
AJ488947.1 | - | 9 | 1462 | |||
AK131261.1 | - | 1 | 51 |