Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MLN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040109.1 | NP_001035198.1 | 2, 3 | 189 | ||
NM_001184698.1 | NP_001171627.1 | 2, 3 | 189 | |||
NM_002418.2 | NP_002409.1 | 2, 3 | 189 | |||
GenBank mRNA | BC069675.1 | - | 2, 3 | 184 | 7262661 | |
BC112314.1 | - | 2, 3 | 153 | 8327782 | ||
BC143981.1 | - | 2, 3 | 153 | 9052499 | ||
Y00695.1 | - | 2, 3 | 173 |