Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MMP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002421.3 | NP_002412.1 | 2 | 297 | ||
GenBank mRNA | AK024818.1 | - | 2 | 220 | ||
AK098450.1 | - | 2 | 220 | |||
AK130687.1 | - | 2 | 220 | |||
AK223035.1 | - | 2 | 178 | |||
AK223281.1 | - | 2 | 220 | |||
AK301340.1 | - | 2 | 223 | |||
BC013875.2 | - | 2 | 196 | 3834572 | ||
BT006874.1 | - | 2 | 154 | |||
DC385245.1 | - | 2 | 297 | |||
M13509.1 | - | 2 | 222 | |||
M15996.1 | - | 2 | 208 | |||
X05231.1 | - | 2 | 220 | |||
X54925.1 | - | 2 | 222 |