Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CACNB2 SIRNA Assays ›
Gene Symbol:
CACNB2
Gene Name:
calcium voltage-gated channel auxiliary subunit beta 2
Gene Aliases:
CACNLB2, CAVB2, MYSB
Chromosome Location:
Chr.10: 18140560 - 18545881 on Build GRCh38
UniGene ID:
Hs.59093
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
114024
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CACNB2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_000724.3
NP_000715.2
9
1472
NM_001167945.1
NP_001161417.1
10
1191
NM_201570.2
NP_963864.1
9
1326
NM_201571.3
NP_963865.2
10
1305
NM_201572.3
NP_963866.2
10
1233
NM_201590.2
NP_963884.2
9
1102
NM_201593.2
NP_963887.2
10
968
NM_201596.2
NP_963890.2
10
1082
NM_201597.2
NP_963891.1
10
1010
XM_005252588.3
XP_005252645.1
9
1216
XM_005252591.3
XP_005252648.1
4
277
XM_006717502.3
XP_006717565.1
9
1135
XM_011519659.2
XP_011517961.1
9
1030
XM_011519660.2
XP_011517962.1
9
1573
XM_017016625.1
XP_016872114.1
10
1190
GenBank mRNA
AB208917.1
-
9
1358
AF137376.1
-
10
1020
AF137377.1
-
9
863
AF285239.2
-
9
860
AF423189.1
-
9
857
AF423190.1
-
10
938
AF423191.1
-
10
1022
AF423192.1
-
9
878
AF465485.1
-
9
752
AK293937.1
-
9
773
AK296471.1
-
10
989
AK299873.1
-
9
1228
AK307526.1
-
10
1191
AK307800.1
-
9
934
AY393858.1
-
10
938
AY393859.1
-
9
860
AY393860.1
-
10
950
AY393861.1
-
10
866
AY393862.1
-
9
788
BC136409.1
-
10
1045
9020397
BM312516.1
-
7
475
S60415.1
-
10
1082
U95019.1
-
9
1358
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_053851, XM_006254304, XM_006254303, XM_006254300, XM_006254301, and others.
Panther Classification:
Molecular Function -
voltage-gated ion channel
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
neuromuscular junction development
visual perception
positive regulation of calcium ion transport
cardiac conduction
calcium ion import
calcium ion transmembrane transport
membrane depolarization during AV node cell action potential
regulation of heart rate by cardiac conduction
establishment of protein localization to plasma membrane
membrane depolarization during atrial cardiac muscle cell action potential
regulation of voltage-gated calcium channel activity
positive regulation of high voltage-gated calcium channel activity
positive regulation of calcium ion transmembrane transport via high voltage-gated calcium channel
voltage-gated calcium channel activity
calcium channel regulator activity
calcium channel activity
protein binding
high voltage-gated calcium channel activity
protein kinase binding
protein domain specific binding
actin filament binding
phosphoprotein binding
voltage-gated calcium channel activity involved in cardiac muscle cell action potential
voltage-gated calcium channel activity involved in AV node cell action potential
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-558c998fd6-nvdhv:80/10.239.163.173:80.
git-commit: a0437b286f8214373ac979cfd76c0df1857d95b8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.14.2-2024.08.97-1.0