Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ITGB2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000211.4 | NP_000202.3 | 16 | 2644 | ||
NM_001127491.2 | NP_001120963.2 | 16 | 2634 | |||
NM_001303238.1 | NP_001290167.1 | 16 | 2599 | |||
XM_006724001.1 | XP_006724064.1 | 16 | 2661 | |||
XM_017028341.1 | XP_016883830.1 | 16 | 2656 | |||
GenBank mRNA | AB208909.1 | - | 15 | 4467 | ||
AK095992.1 | - | 16 | 2599 | |||
AK097216.1 | - | 7 | 1811 | |||
AK097864.1 | - | 1 | 2347 | |||
AK128344.1 | - | 16 | 2469 | |||
AK222505.1 | - | 16 | 2501 | |||
AK303488.1 | - | 16 | 2512 | |||
BC005861.2 | - | 15 | 2455 | 3532902 | ||
BC021077.1 | - | 15 | 2378 | |||
M15395.1 | - | 16 | 2484 |