Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PROS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000313.3 | NP_000304.2 | 2 | 495 | ||
NM_001314077.1 | NP_001301006.1 | 3 | 582 | |||
GenBank mRNA | AK292994.1 | - | 2 | 301 | ||
BC015801.1 | - | 2 | 471 | 3909023 | ||
BG702968.1 | - | 3 | 365 | |||
DA408604.1 | - | 2 | 488 | |||
DB076220.1 | - | 2 | 113 | |||
KU178283.1 | - | 2 | 154 | |||
KU178284.1 | - | 3 | 250 | |||
M14338.1 | - | 2 | 322 | |||
M15036.1 | - | 2 | 300 | |||
X12892.1 | - | 2 | 322 | |||
Y00692.1 | - | 2 | 277 |