Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CRX | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000554.4 | NP_000545.1 | 4 | 1309 | ||
GenBank mRNA | AF335249.1 | - | 1 | 205 | ||
BC016664.1 | - | 4 | 1244 | 3869862 | ||
BC053672.1 | - | 4 | 1195 | 6144191 |