Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRA1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000806.5 | NP_000797.2 | 4 | 560 | ||
NM_001127643.1 | NP_001121115.1 | 4 | 447 | |||
NM_001127644.1 | NP_001121116.1 | 3 | 596 | |||
NM_001127645.1 | NP_001121117.1 | 3 | 184 | |||
NM_001127648.1 | NP_001121120.1 | 2 | 206 | |||
GenBank mRNA | AK223447.1 | - | 3 | 333 | ||
AK289792.1 | - | 4 | 560 | |||
AK295160.1 | - | 4 | 470 | |||
AV720972.1 | - | 3 | 247 | |||
BC030696.1 | - | 3 | 161 | 4820972 | ||
DC307259.1 | - | 3 | 184 | |||
DC325179.1 | - | 3 | 408 | |||
DC334834.1 | - | 2 | 206 | |||
R60404.1 | - | 1 | 20 | |||
X13584.1 | - | 3 | 352 | |||
X14766.1 | - | 3 | 306 |