Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000829.3 | NP_000820.3 | 2, 3 | 531 | ||
NM_001077243.2 | NP_001070711.2 | 2, 3 | 531 | |||
NM_001077244.1 | NP_001070712.1 | 2, 3 | 531 | |||
NM_001112812.1 | NP_001106283.1 | 1, 2 | 399 | |||
NR_046356.1 | 1, 2 | 399 | ||||
XM_005271518.3 | XP_005271575.1 | 2, 3 | 575 | |||
XM_006718823.1 | XP_006718886.1 | 2, 3 | 205 | |||
XM_011542775.2 | XP_011541077.1 | 2, 3 | 575 | |||
XM_017017609.1 | XP_016873098.1 | 2, 3 | 575 | |||
XM_017017610.1 | XP_016873099.1 | 2, 3 | 575 | |||
GenBank mRNA | AB209092.1 | - | 1, 2 | 369 | ||
BC045546.1 | - | 2, 3 | 525 | |||
BC114549.1 | - | 1, 2 | 84 | |||
BC142654.1 | - | 2, 3 | 479 | |||
BC146816.1 | - | 1, 2 | 358 | |||
BC150209.1 | - | 2, 3 | 479 | |||
DA391203.1 | - | 1, 2 | 346 | |||
DB473702.2 | - | 2, 3 | 512 | |||
U16129.1 | - | 1, 2 | 85 |