Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GRIK3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000831.3 | NP_000822.2 | 6 | 977 | ||
GenBank mRNA | AJ249210.1 | - | 6 | 863 | ||
AJ299451.1 | - | 6 | 842 | |||
U16127.1 | - | 6 | 860 |