Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001394.6 | NP_001385.1 | 3, 4 | 1264 | ||
NM_057158.3 | NP_476499.1 | 4, 5 | 2079 | |||
XM_011544428.2 | XP_011542730.1 | 3, 4 | 3824 | |||
GenBank mRNA | AK314820.1 | - | 3, 4 | 1189 | ||
AL137704.1 | - | 4, 5 | 2079 | |||
BC002671.2 | - | 3, 4 | 950 | 3605895 | ||
BC014565.2 | - | 3, 4 | 1182 | 3050199 | ||
BG760517.1 | - | 4, 5 | 472 | |||
S80632.1 | - | 3, 4 | 843 | |||
U21108.1 | - | 3, 4 | 1038 | |||
U48807.1 | - | 3, 4 | 911 |