Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001514.5 | NP_001505.1 | 4 | 450 | ||
XM_011541299.1 | XP_011539601.1 | 4 | 652 | |||
GenBank mRNA | AB451296.1 | - | 4 | 331 | ||
BC020597.1 | - | 4 | 396 | 4807599 | ||
BP379342.1 | - | 4 | 450 | |||
M76766.1 | - | 4 | 369 | |||
S44184.1 | - | 3 | 319 |