Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other INPP4A SIRNA Assays ›
Gene Symbol:
INPP4A
Gene Name:
inositol polyphosphate-4-phosphatase type I A
Gene Aliases:
INPP4, TVAS1
Chromosome Location:
Chr.2: 98444592 - 98594068 on Build GRCh38
UniGene ID:
Hs.469386
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
114164
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: INPP4A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001134224.1
NP_001127696.1
10
1111
NM_001134225.1
NP_001127697.1
10
1111
NM_001566.2
NP_001557.1
10
1111
NM_004027.2
NP_004018.1
10
1111
XM_006712499.3
XP_006712562.1
10
1106
XM_006712500.3
XP_006712563.1
10
1106
XM_006712501.3
XP_006712564.1
10
1106
XM_006712502.3
XP_006712565.1
10
1106
XM_006712503.3
XP_006712566.1
10
1106
XM_006712504.3
XP_006712567.1
10
1106
XM_006712505.3
XP_006712568.1
10
1106
XM_006712506.3
XP_006712569.1
10
1106
XM_006712507.3
XP_006712570.1
10
1106
XM_006712508.3
XP_006712571.1
10
1106
XM_011511122.2
XP_011509424.1
9
1056
XM_011511123.2
XP_011509425.1
10
1106
XM_011511124.2
XP_011509426.1
10
1106
XM_011511125.2
XP_011509427.1
7
988
XM_017003993.1
XP_016859482.1
10
947
XM_017003994.1
XP_016859483.1
10
1106
XM_017003995.1
XP_016859484.1
10
1106
XM_017003996.1
XP_016859485.1
10
1106
XM_017003997.1
XP_016859486.1
10
1165
XM_017003998.1
XP_016859487.1
10
1165
XM_017003999.1
XP_016859488.1
10
1164
XM_017004000.1
XP_016859489.1
10
1106
XM_017004001.1
XP_016859490.1
10
1106
XM_017004002.1
XP_016859491.1
10
1024
XM_017004003.1
XP_016859492.1
10
1106
GenBank mRNA
AF368319.1
-
8
718
BC028361.2
-
10
1111
4830663
U26398.1
-
10
1012
U96919.1
-
10
1012
Back To Top
More Information
Gene Ontology Categories:
Function(s)
Process(es)
phosphatidylinositol biosynthetic process
signal transduction
dephosphorylation
phosphatidylinositol-3-phosphate biosynthetic process
inositol phosphate metabolic process
protein binding
phosphatidylinositol-3,4-bisphosphate 4-phosphatase activity
inositol-1,3,4-trisphosphate 4-phosphatase activity
phosphatidylinositol-4,5-bisphosphate 4-phosphatase activity
inositol-3,4-bisphosphate 4-phosphatase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-jlg4l:80/100.66.77.222:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline