Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALPI | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001631.4 | NP_001622.2 | 10 | 1312 | ||
GenBank mRNA | AK313163.1 | - | 10 | 1301 | ||
BC132678.1 | - | 10 | 1254 | 40146700 | ||
M15694.1 | - | 10 | 1224 |