Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPG | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252296.1 | NP_001239225.1 | 2 | 1123 | ||
NM_001806.3 | NP_001797.1 | 2 | 1260 | |||
GenBank mRNA | BC013128.1 | - | 2 | 1178 | 3445301 |