Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001938.2 | NP_001929.1 | 1, 2 | 940 | ||
GenBank mRNA | AK057541.1 | - | 2, 3 | 802 | ||
AK223442.1 | - | 1, 2 | 954 | |||
AL833729.1 | - | 1, 2 | 923 | |||
BC002809.2 | - | 1, 2 | 917 | 3634908 | ||
BC035507.2 | - | 1, 2 | 940 | 4555131 | ||
BC068553.1 | - | 1, 2 | 939 | 4822070 | ||
BT006972.1 | - | 1, 2 | 215 | |||
M97388.1 | - | 2, 3 | 762 |