Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GABRR1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256703.1 | NP_001243632.1 | 2 | 643 | ||
NM_001256704.1 | NP_001243633.1 | 4 | 796 | |||
NM_001267582.1 | NP_001254511.1 | 5 | 464 | |||
NM_002042.4 | NP_002033.2 | 3 | 694 | |||
XM_017010689.1 | XP_016866178.1 | 4 | 284 | |||
GenBank mRNA | AK296124.1 | - | 2 | 235 | ||
AK298948.1 | - | 5 | 454 | |||
AK313609.1 | - | 4 | 796 | |||
BC130344.1 | - | 3 | 248 | 40146375 | ||
DA753437.1 | - | 4 | 390 | |||
M62400.1 | - | 3 | 267 |