Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002146.4 | NP_002137.4 | 3 | 1237 | ||
XM_005257277.3 | XP_005257334.1 | 4 | 951 | |||
XM_005257280.3 | XP_005257337.1 | 4 | 862 | |||
XM_006721854.2 | XP_006721917.1 | 4 | 969 | |||
XM_011524707.2 | XP_011523009.1 | 4 | 10777 | |||
XM_011524710.1 | XP_011523012.1 | 4 | 3177 | |||
XM_011524719.1 | XP_011523021.1 | 3 | 5658 | |||
XM_011524720.2 | XP_011523022.1 | 3 | 8349 | |||
XM_011524721.2 | XP_011523023.1 | 3 | 3484 | |||
XM_011524726.2 | XP_011523028.1 | 3 | 435 | |||
XM_017024557.1 | XP_016880046.1 | 4 | 10525 | |||
XM_017024558.1 | XP_016880047.1 | 4 | 10399 | |||
XM_017024559.1 | XP_016880048.1 | 4 | 10241 | |||
XM_017024560.1 | XP_016880049.1 | 4 | 10485 | |||
XM_017024561.1 | XP_016880050.1 | 4 | 14322 | |||
XM_017024562.1 | XP_016880051.1 | 4 | 10263 | |||
XM_017024563.1 | XP_016880052.1 | 5 | 12918 | |||
GenBank mRNA | AK027609.1 | - | 3 | 1275 | ||
AK291182.1 | - | 4 | 948 | |||
AK299265.1 | - | 4 | 694 | |||
AK315102.1 | - | 2 | 503 | |||
AK316245.1 | - | 3 | 470 | |||
U59298.1 | - | 1 | 1828 | |||
X16667.1 | - | 3 | 742 |