Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002146.4 | NP_002137.4 | 4 | 2541 | ||
XM_005257277.3 | XP_005257334.1 | 5 | 2255 | |||
XM_005257280.3 | XP_005257337.1 | 5 | 2166 | |||
XM_005257282.3 | XP_005257339.1 | 5 | 1731 | |||
XM_006721854.2 | XP_006721917.1 | 5 | 2273 | |||
XM_011524707.2 | XP_011523009.1 | 5 | 12081 | |||
XM_011524708.2 | XP_011523010.1 | 4 | 11661 | |||
XM_011524710.1 | XP_011523012.1 | 5 | 4481 | |||
XM_011524719.1 | XP_011523021.1 | 4 | 6962 | |||
XM_011524720.2 | XP_011523022.1 | 4 | 9653 | |||
XM_011524721.2 | XP_011523023.1 | 4 | 4785 | |||
XM_011524726.2 | XP_011523028.1 | 4 | 1739 | |||
XM_017024557.1 | XP_016880046.1 | 5 | 11829 | |||
XM_017024558.1 | XP_016880047.1 | 5 | 11703 | |||
XM_017024559.1 | XP_016880048.1 | 5 | 11545 | |||
XM_017024560.1 | XP_016880049.1 | 5 | 11789 | |||
XM_017024561.1 | XP_016880050.1 | 5 | 15626 | |||
XM_017024562.1 | XP_016880051.1 | 5 | 11567 | |||
XM_017024563.1 | XP_016880052.1 | 6 | 14222 | |||
GenBank mRNA | AK027609.1 | - | 4 | 2579 | ||
AK316245.1 | - | 4 | 1774 |