Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MEIS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002398.2 | NP_002389.1 | 6 | 954 | ||
XM_005264321.2 | XP_005264378.1 | 7 | 1277 | |||
XM_005264322.2 | XP_005264379.1 | 6 | 1229 | |||
XM_005264323.2 | XP_005264380.1 | 7 | 1277 | |||
XM_005264324.4 | XP_005264381.1 | 5 | 350 | |||
XM_005264325.4 | XP_005264382.1 | 5 | 1960 | |||
XM_017004141.1 | XP_016859630.1 | 7 | 1277 | |||
XM_017004142.1 | XP_016859631.1 | 6 | 1229 | |||
XM_017004143.1 | XP_016859632.1 | 5 | 1960 | |||
XM_017004144.1 | XP_016859633.1 | 6 | 1135 | |||
XM_017004145.1 | XP_016859634.1 | 5 | 1087 | |||
XM_017004146.1 | XP_016859635.1 | 5 | 1087 | |||
GenBank mRNA | AK098174.1 | - | 4 | 331 | ||
AK297109.1 | - | 5 | 609 | |||
AK297817.1 | - | 5 | 415 | |||
AK314949.1 | - | 6 | 704 | |||
AL832770.1 | - | 6 | 967 | |||
BC043503.1 | - | 6 | 949 | 5531644 | ||
U85707.1 | - | 6 | 562 |