Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LDHB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001174097.2 | NP_001167568.1 | 6 | 966 | ||
NM_001315537.1 | NP_001302466.1 | 6 | 744 | |||
NM_002300.7 | NP_002291.1 | 6 | 744 | |||
XM_006719074.2 | XP_006719137.1 | 6 | 860 | |||
GenBank mRNA | AB209231.1 | - | 1 | 1166 | ||
AK129881.1 | - | 6 | 695 | |||
AK312328.1 | - | 6 | 728 | |||
AL516910.3 | - | 6 | 965 | |||
AY189689.1 | - | 6 | 720 | |||
BC002362.1 | - | 6 | 711 | 2961445 | ||
BC015122.1 | - | 6 | 735 | 3685985 | ||
BC071860.1 | - | 6 | 676 | 4902461 | ||
BT019764.1 | - | 5 | 633 | |||
BT019765.1 | - | 5 | 633 | |||
EU794633.1 | - | 6 | 687 | |||
Y00711.1 | - | 6 | 687 |