Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NPAS2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002518.3 | NP_002509.2 | 3 | 431 | ||
XM_005263953.2 | XP_005264010.1 | 3 | 342 | |||
XM_005263959.2 | XP_005264016.1 | 3 | 342 | |||
XM_005263960.2 | XP_005264017.1 | 3 | 342 | |||
XM_011511242.2 | XP_011509544.1 | 3 | 399 | |||
XM_011511243.2 | XP_011509545.1 | 3 | 342 | |||
XM_017004214.1 | XP_016859703.1 | 3 | 342 | |||
XM_017004215.1 | XP_016859704.1 | 3 | 342 | |||
XM_017004216.1 | XP_016859705.1 | 3 | 342 | |||
XM_017004217.1 | XP_016859706.1 | 3 | 342 | |||
GenBank mRNA | AK294733.1 | - | 3 | 342 | ||
BC028107.1 | - | 2 | 174 | |||
BC051351.1 | - | 3 | 395 | 6538324 | ||
BC072383.1 | - | 3 | 398 | 6371187 | ||
KJ534892.1 | - | 2 | 145 | |||
U77970.1 | - | 3 | 431 |