Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POLR2G | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002696.2 | NP_002687.1 | 3 | 267 | ||
GenBank mRNA | AK312131.1 | - | 3 | 226 | ||
BC112162.1 | - | 3 | 206 | 8327630 | ||
BC112164.1 | - | 3 | 206 | 8327632 | ||
BG530809.1 | - | 3 | 284 | |||
U20659.1 | - | 3 | 185 |