Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190447.1 | NP_001177376.1 | 13 | 4128 | ||
NM_002718.4 | NP_002709.2 | 14 | 6563 | |||
NM_181897.2 | NP_871626.1 | 13 | 4418 | |||
XM_006713686.3 | XP_006713749.1 | 14 | 6974 | |||
XM_011512956.2 | XP_011511258.1 | 14 | 6738 | |||
XM_017006787.1 | XP_016862276.1 | 13 | 6296 | |||
GenBank mRNA | AL389975.1 | - | 11 | 3646 |