Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUPT4H1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003168.2 | NP_003159.1 | 5 | 596 | ||
NR_073470.1 | 5 | 598 | ||||
GenBank mRNA | BC002802.2 | - | 5 | 554 | 3636217 | |
BP318625.1 | - | 5 | 559 | |||
BX331655.2 | - | 5 | 560 | |||
U38817.1 | - | 5 | 535 | |||
U38818.1 | - | 5 | 554 | |||
U43923.1 | - | 5 | 560 |