Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TFAP4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003223.2 | NP_003214.1 | 7 | 2102 | ||
XM_011522633.2 | XP_011520935.1 | 7 | 2238 | |||
XM_011522635.2 | XP_011520937.1 | 7 | 1918 | |||
GenBank mRNA | AB209146.1 | - | 5 | 6452 | ||
BC010576.1 | - | 7 | 1990 | 4181538 | ||
S73885.1 | - | 7 | 2104 | |||
X57435.1 | - | 7 | 1795 |