Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: USF2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321150.1 | NP_001308079.1 | 4, 5 | 396 | ||
NM_003367.3 | NP_003358.1 | 6, 7 | 789 | |||
NM_207291.2 | NP_997174.1 | 5, 6 | 588 | |||
XM_005259197.3 | XP_005259254.1 | 6, 7 | 688 | |||
XM_011527260.2 | XP_011525562.1 | 6, 7 | 688 | |||
XM_011527261.2 | XP_011525563.1 | 6, 7 | 688 | |||
XM_017027199.1 | XP_016882688.1 | 5, 6 | 487 | |||
GenBank mRNA | AK297022.1 | - | 6, 7 | 659 | ||
AY147880.1 | - | 4, 5 | 272 | |||
BC049821.1 | - | 6, 7 | 702 | 5483523 | ||
CR536504.1 | - | 6, 7 | 665 | |||
M77476.1 | - | 3, 4 | 389 | |||
S50537.1 | - | 2, 3 | 329 | |||
X90824.1 | - | 4, 5 | 528 | |||
X90825.1 | - | 5, 6 | 573 | |||
X90826.1 | - | 6, 7 | 671 |