Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: XPO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003400.3 | NP_003391.1 | 2, 3 | 862 | ||
XM_005264544.1 | XP_005264601.1 | 2, 3 | 884 | |||
XM_006712094.2 | XP_006712157.1 | 3, 4 | 1005 | |||
XM_011533097.1 | XP_011531399.1 | 2, 3 | 171 | |||
XM_011533098.1 | XP_011531400.1 | 2, 3 | 839 | |||
XM_011533099.2 | XP_011531401.1 | 2, 3 | 884 | |||
GenBank mRNA | AK289920.1 | - | 2, 3 | 541 | ||
AK312426.1 | - | 2, 3 | 300 | |||
AL833550.1 | - | 2, 3 | 809 | |||
BC032847.2 | - | 3, 4 | 263 | 5267242 | ||
BX647758.1 | - | 2, 3 | 844 | |||
CR749840.1 | - | 3, 4 | 636 | |||
D89729.1 | - | 1, 2 | 152 | |||
Y08614.2 | - | 2, 3 | 172 |