Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAX8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003466.3 | NP_003457.1 | 12 | 2137 | ||
NM_013952.3 | NP_039246.1 | 12 | 2058 | |||
NM_013953.3 | NP_039247.1 | 10 | 1827 | |||
NM_013992.3 | NP_054698.1 | 9 | 1725 | |||
XM_011511790.1 | XP_011510092.1 | 12 | 2169 | |||
XM_011511791.1 | XP_011510093.1 | 12 | 2225 | |||
XM_011511792.1 | XP_011510094.1 | 12 | 2090 | |||
XM_011511793.1 | XP_011510095.1 | 11 | 1980 | |||
XM_011511794.1 | XP_011510096.1 | 11 | 1949 | |||
GenBank mRNA | AK096795.1 | - | 1 | 1647 | ||
AK225538.1 | - | 12 | 1968 | |||
AK307173.1 | - | 4 | 975 | |||
BC001060.2 | - | 12 | 2128 | 2963877 | ||
CR936798.1 | - | 1 | 7622 | |||
KC733810.1 | - | 12 | 2234 | |||
X69699.1 | - | 12 | 2127 |