Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DUSP11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_003584.2 | NP_003575.2 | 7 | 785 | ||
GenBank mRNA | AF023917.1 | - | 7 | 726 | ||
AK315271.1 | - | 7 | 737 | |||
BC000346.2 | - | 7 | 703 | 2960939 |