Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRP6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002336.2 | NP_002327.2 | 4 | 814 | ||
XM_006719078.3 | XP_006719141.1 | 4 | 958 | |||
XM_011520671.2 | XP_011518973.1 | 3 | 564 | |||
XM_017019304.1 | XP_016874793.1 | 2 | 632 | |||
GenBank mRNA | AB209683.1 | - | 3 | 266 | ||
AF074264.1 | - | 4 | 749 | |||
BC117136.1 | - | 4 | 706 | 40125687 | ||
BC126405.1 | - | 4 | 706 | 8992121 | ||
BC143725.1 | - | 4 | 706 | 9052239 |