Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ASCL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004316.3 | NP_004307.2 | 2 | 1645 | ||
GenBank mRNA | BC001638.2 | - | 2 | 1632 | ||
BC004425.1 | - | 2 | 1632 | 2819879 | ||
CF454566.1 | - | 2 | 402 | |||
L08424.1 | - | 2 | 1511 |