Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HOXC6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004503.3 | NP_004494.1 | 2 | 964 | ||
NM_153693.4 | NP_710160.1 | 3 | 1361 | |||
GenBank mRNA | BF058345.1 | - | 1 | 425 | ||
M16938.1 | - | 3 | 1088 | |||
S82986.1 | - | 2 | 984 |