Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PKNOX1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001320694.1 | NP_001307623.1 | 3, 4 | 384 | ||
NM_004571.4 | NP_004562.2 | 3, 4 | 384 | |||
GenBank mRNA | AK222892.1 | - | 2, 3 | 250 | ||
AY142115.1 | - | 3, 4 | 361 | |||
AY196965.1 | - | 3, 4 | 362 | |||
AY566249.1 | - | 2, 3 | 232 | |||
BC000735.2 | - | 3, 4 | 330 | |||
BC007746.2 | - | 2, 3 | 191 | 4099156 | ||
BP369805.1 | - | 3, 4 | 384 | |||
DR000771.1 | - | 3, 4 | 311 | |||
U68727.1 | - | 2, 3 | 255 | |||
Y13613.1 | - | 3, 4 | 297 |