Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RAB4A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004578.3 | NP_004569.2 | 5 | 585 | ||
GenBank mRNA | AF498934.1 | - | 5 | 307 | ||
AK223009.1 | - | 5 | 350 | |||
AK313807.1 | - | 5 | 558 | |||
AY585832.1 | - | 5 | 514 | |||
BC002438.1 | - | 5 | 378 | 3346455 | ||
BC004309.1 | - | 5 | 515 | 3627110 | ||
M28211.1 | - | 5 | 362 | |||
X13902.1 | - | 5 | 418 |