Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MTMR4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004687.4 | NP_004678.3 | 16 | 2416 | ||
XM_005257784.2 | XP_005257841.1 | 15 | 2565 | |||
XM_005257785.4 | XP_005257842.1 | 17 | 2533 | |||
XM_005257786.4 | XP_005257843.1 | 16 | 3320 | |||
XM_006722168.3 | XP_006722231.1 | 16 | 2360 | |||
XM_011525460.2 | XP_011523762.1 | 17 | 3413 | |||
GenBank mRNA | AB014547.2 | - | 14 | 2306 | ||
AF264717.1 | - | 16 | 2359 | |||
BC035609.1 | - | 16 | 2377 | 5201989 |