Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CTDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001202504.1 | NP_001189433.1 | 13 | 2650 | ||
NM_001318511.1 | NP_001305440.1 | 12 | 2856 | |||
NM_004715.4 | NP_004706.3 | 13 | 3023 | |||
NM_048368.3 | NP_430255.2 | 12 | 2860 | |||
XM_011526261.1 | XP_011524563.1 | 11 | 2671 | |||
XM_017026078.1 | XP_016881567.1 | 11 | 2503 | |||
GenBank mRNA | AF081287.1 | - | 12 | 2571 | ||
AF154115.1 | - | 13 | 2959 | |||
AK123180.1 | - | 2 | 1041 | |||
BC015010.1 | - | 6 | 1807 | |||
BC032515.1 | - | 12 | 2510 | |||
BC047999.1 | - | 13 | 3023 | |||
BC052576.1 | - | 13 | 2615 | |||
BC063447.1 | - | 12 | 2815 | 5500032 | ||
BM512113.1 | - | 1 | 103 | |||
BX103692.1 | - | 1 | 99 |