Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MSC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005098.3 | NP_005089.2 | 1, 2 | 857 | ||
GenBank mRNA | AF060154.1 | - | 1, 2 | 751 | ||
AF087036.1 | - | 1, 2 | 525 | |||
AK313102.1 | - | 1, 2 | 814 | |||
BC006313.2 | - | 1, 2 | 823 | 4123825 | ||
BC067827.1 | - | 1, 2 | 832 | 30336424 | ||
BT007169.1 | - | 1, 2 | 539 |