Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005413.3 | NP_005404.1 | 2 | 1192 | ||
GenBank mRNA | AJ012611.1 | - | 2 | 1192 | ||
BF569811.1 | - | 2 | 495 |