Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SIX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005413.3 | NP_005404.1 | 2 | 1330 | ||
GenBank mRNA | AJ012611.1 | - | 2 | 1330 | ||
EL952320.1 | - | 1 | 55 |