Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284209.1 | NP_001271138.1 | 4 | 386 | ||
NM_001284210.1 | NP_001271139.1 | 4 | 386 | |||
NM_001284211.1 | NP_001271140.1 | 4 | 386 | |||
NM_005466.3 | NP_005457.2 | 4 | 386 | |||
GenBank mRNA | AF074723.1 | - | 4 | 357 | ||
AF155104.1 | - | 3 | 308 | |||
AK222587.1 | - | 4 | 362 | |||
AK223346.1 | - | 4 | 351 | |||
AK290647.1 | - | 4 | 351 | |||
AK300417.1 | - | 4 | 351 | |||
AK300460.1 | - | 4 | 351 | |||
AK304361.1 | - | 4 | 351 | |||
BC004106.2 | - | 4 | 360 | 3936675 | ||
BM152825.1 | - | 4 | 376 | |||
BM802752.1 | - | 338 | ||||
BT006831.1 | - | 4 | 333 | |||
CR541697.1 | - | 4 | 333 | |||
HY064772.1 | - | 4 | 387 | |||
U78082.1 | - | 4 | 383 |