Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MED6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284209.1 | NP_001271138.1 | 6 | 608 | ||
NM_001284211.1 | NP_001271140.1 | 6 | 587 | |||
NM_005466.3 | NP_005457.2 | 6 | 587 | |||
GenBank mRNA | AF074723.1 | - | 6 | 558 | ||
AF155104.1 | - | 5 | 509 | |||
AK222587.1 | - | 6 | 563 | |||
AK223346.1 | - | 6 | 552 | |||
AK290647.1 | - | 6 | 552 | |||
AK300460.1 | - | 6 | 573 | |||
BC004106.2 | - | 6 | 561 | 3936675 | ||
BM802752.1 | - | 539 | ||||
BT006831.1 | - | 6 | 534 | |||
CR541697.1 | - | 6 | 534 |