Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTF2E1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005513.2 | NP_005504.2 | 5 | 2317 | ||
XM_011512744.2 | XP_011511046.1 | 5 | 2440 | |||
XM_011512745.2 | XP_011511047.1 | 5 | 2388 | |||
GenBank mRNA | AK223401.1 | - | 6 | 1661 | ||
X63468.1 | - | 4 | 2257 |