Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LYL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005583.4 | NP_005574.2 | 2 | 834 | ||
XM_005259912.3 | XP_005259969.1 | 2 | 672 | |||
XM_017026816.1 | XP_016882305.1 | 3 | 1534 | |||
XM_017026817.1 | XP_016882306.1 | 3 | 1387 | |||
GenBank mRNA | BC002796.2 | - | 2 | 541 | 3619444 | |
M22637.1 | - | 2 | 399 |