Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPRY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001258038.1 | NP_001244967.1 | 3 | 613 | ||
NM_001258039.1 | NP_001244968.1 | 2 | 357 | |||
NM_005841.2 | NP_005832.1 | 2 | 534 | |||
NM_199327.2 | NP_955359.1 | 2 | 367 | |||
XM_005262686.1 | XP_005262743.1 | 3 | 1274 | |||
XM_017007635.1 | XP_016863124.1 | 2 | 347 | |||
GenBank mRNA | AK026960.1 | - | 2 | 514 | ||
AY192146.1 | - | 2 | 362 | |||
AY590694.1 | - | 1 | 165 | |||
BC063856.1 | - | 2 | 496 | 6181674 | ||
CN302060.1 | - | 2 | 337 | |||
DB250100.1 | - | 2 | 534 |