Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MXI1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001008541.1 | NP_001008541.1 | 5 | 1238 | ||
NM_005962.4 | NP_005953.4 | 6 | 1463 | |||
NM_130439.3 | NP_569157.2 | 6 | 1661 | |||
GenBank mRNA | AK309718.1 | - | 6 | 1273 | ||
BC012907.1 | - | 5 | 1234 | 3882557 | ||
BC035128.1 | - | 6 | 1662 | 5263647 |