Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MAGEB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_002365.4 | NP_002356.2 | 5 | 1228 | ||
XM_011545513.2 | XP_011543815.1 | 4 | 835 | |||
XM_011545514.2 | XP_011543816.1 | 3 | 802 | |||
GenBank mRNA | BC069784.1 | - | 1 | 550 | 7262601 |