Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HDAC4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006037.3 | NP_006028.2 | 7, 8 | 1533 | ||
XM_006712877.3 | XP_006712940.1 | 6, 7 | 7479 | |||
XM_006712878.3 | XP_006712941.1 | 7, 8 | 1038 | |||
XM_006712879.3 | XP_006712942.1 | 6, 7 | 6931 | |||
XM_006712880.3 | XP_006712943.1 | 6, 7 | 6943 | |||
XM_011512217.2 | XP_011510519.1 | 6, 7 | 7479 | |||
XM_011512218.2 | XP_011510520.1 | 6, 7 | 7479 | |||
XM_011512219.2 | XP_011510521.1 | 6, 7 | 7479 | |||
XM_011512220.2 | XP_011510522.1 | 6, 7 | 1218 | |||
XM_011512221.1 | XP_011510523.1 | 7, 8 | 1206 | |||
XM_011512222.2 | XP_011510524.1 | 7, 8 | 1507 | |||
XM_011512223.2 | XP_011510525.1 | 7, 8 | 1021 | |||
XM_011512224.2 | XP_011510526.1 | 6, 7 | 1409 | |||
XM_011512225.2 | XP_011510527.1 | 6, 7 | 1409 | |||
XM_011512226.2 | XP_011510528.1 | 6, 7 | 966 | |||
XM_011512227.2 | XP_011510529.1 | 5, 6 | 6493 | |||
XM_017005395.1 | XP_016860884.1 | 3, 4 | 270 | |||
GenBank mRNA | AB006626.2 | - | 7, 8 | 1533 | ||
AF132607.1 | - | 6, 7 | 741 | |||
AK303368.1 | - | 6, 7 | 1085 | |||
AK303467.1 | - | 5, 6 | 725 | |||
AK303617.1 | - | 4, 5 | 714 | |||
AK308791.1 | - | 7, 8 | 1211 | |||
AK310603.1 | - | 6, 7 | 1193 | |||
BC039904.1 | - | 7, 8 | 785 | 6067363 |