Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ENPP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040092.2 | NP_001035181.1 | 7 | 723 | ||
NM_001130863.2 | NP_001124335.1 | 7 | 723 | |||
NM_006209.4 | NP_006200.3 | 7 | 723 | |||
XM_006716584.1 | XP_006716647.1 | 7 | 701 | |||
XM_006716585.1 | XP_006716648.1 | 7 | 701 | |||
XM_006716587.1 | XP_006716650.1 | 7 | 701 | |||
XM_017013571.1 | XP_016869060.1 | 7 | 757 | |||
XM_017013572.1 | XP_016869061.1 | 7 | 701 | |||
XM_017013573.1 | XP_016869062.1 | 7 | 757 | |||
XM_017013574.1 | XP_016869063.1 | 7 | 757 | |||
XM_017013575.1 | XP_016869064.1 | 7 | 701 | |||
GenBank mRNA | AK315253.1 | - | 7 | 696 | ||
BC034961.2 | - | 7 | 696 | 4828484 | ||
D45421.1 | - | 7 | 670 | |||
EU131011.2 | - | 7 | 637 | |||
L35594.1 | - | 7 | 686 |