Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R5E | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001282179.1 | NP_001269108.1 | 2 | 718 | ||
NM_001282180.1 | NP_001269109.1 | 2 | 721 | |||
NM_006246.3 | NP_006237.1 | 2 | 721 | |||
NR_104104.1 | 2 | 721 | ||||
GenBank mRNA | AB209811.1 | - | 2 | 674 | ||
AK299520.1 | - | 2 | 658 | |||
AK313500.1 | - | 2 | 574 | |||
BC013154.1 | - | 2 | 714 | |||
BC034994.1 | - | 3 | 715 | |||
BC048305.1 | - | 2 | 454 | |||
BC064358.1 | - | 2 | 701 | |||
BC073873.1 | - | 2 | 673 | |||
BC092477.1 | - | 2 | 691 | 5267163 | ||
BC093766.1 | - | 2 | 189 | 7939611 | ||
BC101479.1 | - | 2 | 189 | 8068985 | ||
BC143231.1 | - | 2 | 186 | 9051732 | ||
BC143234.1 | - | 2 | 189 | 9051735 | ||
L76703.1 | - | 2 | 698 | |||
Z69029.1 | - | 2 | 636 |